missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) Chloride Hexahydrate, ≥98%, Reagent Grade, Honeywell™

Product Code. 15667860
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
Packaging:
Poly bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade, chunks,≥98%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-77-1
Molecular Formula Cl3FeH12O6
Molecular Weight (g/mol) 270.29
MDL Number MFCD00149712
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
IUPAC Name iron(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Boiling Point 280°C to 285°C
Melting Point 37°C
Color Brown
pH 1.8 (at 10.00g/L 25°C)
Physical Form Solid
Quantity 1 kg
Linear Formula FeCl3 · 6H2O
UN Number UN3260
Formula Weight 270.3g/mol
Percent Purity ≥98%
Odor Weak
Packaging Poly bottle
Vapor Pressure 1mmHg (194°C)
Density 2.8g/cu.cm at 20°C
Chemical Name or Material Iron(III) chloride hexahydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.