missing translation for 'onlineSavingsMsg'
Learn More
Iron(III) chloride hexahydrate, 99+%, for analysis, Thermo Scientific Chemicals Product Code.: 10695862

Iron(III) chloride hexahydrate, 99+%, for analysis, Thermo Scientific Chemicals

Product Code. 10695862
100 g, Plastic bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10695862

Brand: Thermo Scientific Acros 217091000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid for analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-77-1
Molecular Formula Cl3FeH12O6
Molecular Weight (g/mol) 270.29
MDL Number MFCD00149712
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
Synonym Ferric chloride hexahydrate
IUPAC Name iron(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Boiling Point 280.0°C to 285.0°C
Melting Point 37.0°C
Color Orange
Physical Form Adhering Crystalline Powder and Lumps
Quantity 100 g
Assay Percent Range 99.0 to 102% (Iodometry)
Linear Formula FeCl3·6H2O
Fieser 01,390; 02,199; 03,145; 04,236; 05,307; 06,259; 07,153; 08,228; 09,222; 15,158
Merck Index 15, 4046
Solubility Information Solubility in water: 920g/L (20°C). Other solubilities: readily soluble in alcohol, acetone and ether
Formula Weight 270.29
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Iron(III) chloride hexahydrate, p.a.
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467796 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number217091000,217090025,217095000
Lot NumberA0467795 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number217091000,217090025,217095000
Lot NumberA0450173 Certificate TypeCertificate of Analysis Date13/03/2024 Catalog Number217091000,217090025,217095000
Lot NumberA0460688 Certificate TypeCertificate of Analysis Date28/02/2024 Catalog Number217091000,217090025,217095000
Lot NumberA0445029 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number217091000,217090025,217095000
compliance-icons compliance-icons
Product Identifier
  • Iron(III) chloride hexahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Iron (III) chloridehexahydrate, Iron(III) chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iron(III) chloride hexahydrate, 99+%, for analysis, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.