missing translation for 'onlineSavingsMsg'
Learn More

Iron(Iii) Chloride Hexahydrate, ACS Reagent, Honeywell Fluka™

ACS Reagent, 97%

£20.09 - £783.60

Chemical Identifiers

CAS 10025-77-1
Molecular Formula Cl3FeH12O6
Molecular Weight (g/mol) 270.29
MDL Number MFCD00149712
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
IUPAC Name iron(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15602340
View Documents
Honeywell-Fluka
236489-5G
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15682330
View Documents
Honeywell-Fluka
236489-100G
100 g
£49.69
100g
Please to purchase this item. Need a web account? Register with us today!
15692330
View Documents
Honeywell-Fluka
236489-500G
500 g
£150.30
500g
Please to purchase this item. Need a web account? Register with us today!
15612340
View Documents
Honeywell-Fluka
236489-6X500G
6 x 500 g
£783.60
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

10025-77-1
270.29
NQXWGWZJXJUMQB-UHFFFAOYSA-K
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]
Cl3FeH12O6
MFCD00149712
iron(3+) hexahydrate trichloride

Specifications

10025-77-1
FeCl3 · 6H2O
3260
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]
270.29
97%
1mmHg (194°C)
Cl3FeH12O6
MFCD00149712
NQXWGWZJXJUMQB-UHFFFAOYSA-K
iron(3+) hexahydrate trichloride
270.3g/mol
ACS Reagent
Iron(Iii) Chloride Hexahydrate
Videos
Safety and Handling

Safety and Handling

P280-P305 + P351 + P338

H290-H302-H315-H318

missing translation for 'einecsNumber' : 231-729-4

missing translation for 'rtecsNumber' : NO5425000

SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.