missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(III) Chloride Hexahydrate, Extra Pure, SLR, Fisher Chemical™
80.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
1 kg
5 kg
Specifications
Boiling Point | 280°C |
Melting Point | 37°C |
Color | Yellow |
pH | 2 |
Physical Form | Solid |
Quantity | 1 kg |
Formula Weight | 270.29g/mol |
Packaging | Amber glass bottle |
Safety and Handling
Product Identifier
- Iron(III) chloride hexahydrate
- Danger
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Serious eye damage/eye irritation Category 1
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H317-May cause an allergic skin reaction.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P313-Get medical advice/attention.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Iron(III) chloride,Iron (III) chloridehexahydrate
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'