missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(III) chloride hexahydrate, Honeywell Fluka™
Click to view available options
Quantity:
50 g
250 g
1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 50 g |
Linear Formula | FeCl3 · 6H2O |
UN Number | UN3260 |
Formula Weight | 270.3g/mol |
Percent Purity | 98 to 102% (RT) |
Vapor Pressure | 1 mmHg (194°C) |
Chemical Name or Material | Iron(III) chloride hexahydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'