missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(Iii) Chloride Hexahydrate, puriss. p.a., Honeywell Fluka™
Click to view available options
Quantity:
250 g
1 kg
25 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 25 kg |
Linear Formula | FeCl3 · 6H2O |
UN Number | 3260 |
Formula Weight | 270.3g/mol |
Percent Purity | ≥99% |
Grade | puriss. p.a. |
Vapor Pressure | 1mmHg (194°C) |
Chemical Name or Material | Iron(Iii) Chloride Hexahydrate, Reag. Ph. Eur. |
Safety and Handling
Product Identifier
- Iron(III) chloride hexahydrate
- Danger
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- P234-Keep only in original container.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'