Learn More
Iron(III) hydroxide, gamma-phase, Thermo Scientific Chemicals
Description
Iron(III) hydroxide, γ-phase is used in purifying water and as an absorbent in chemical processes. It is involved in the preparation of iron oxide-hydroxides nanoparticles, which is used as a very good adsorbent for lead removal from aquatic media. It is also used as a pigment, as a catalyst and in aquarium water treatment as a phosphate binder. Further, it is employed in the paints-lacquers-varnishes industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 135°C (decomposition) |
Physical Form | Powder |
Quantity | 2 g |
Merck Index | 14,4025 |
Solubility Information | Insoluble in water. |
Formula Weight | 88.85 |
Odor | Odorless |
Chemical Name or Material | Iron(III) hydroxide, gamma-phase |
RUO – Research Use Only