missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(III) isopropoxide, 2.5% w/v in isopropanol, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 mL
25 mL
Description
Iron(III) isopropoxide is used in nano-materials & thin films, engineering & environmental, catalysis & synthesis and energy conversion & storage.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Red to Brown |
Density | 0.79 g/mL |
Boiling Point | 82°C |
Flash Point | 12°C (54°F) |
Odor | Alcohol-like |
Quantity | 25 mL |
UN Number | UN1219 |
Sensitivity | Moisture sensitive |
Solubility Information | It hydrolyzes with water. |
Formula Weight | 233.11 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'