missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) nitrate nonahydrate, 99+%, for analysis, Thermo Scientific Chemicals

Product Code. 11934271
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-61-8
Molecular Formula FeH18N3O18
Molecular Weight (g/mol) 403.99
MDL Number MFCD00149708
InChI Key SZQUEWJRBJDHSM-UHFFFAOYSA-N
Synonym ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate
PubChem CID 16211566
SMILES O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 47.0°C
Color Purple
Physical Form Slightly Adhering Crystalline Powder
Quantity 5 kg
Assay Percent Range 99% min. (Iodometry)
Linear Formula Fe(NO3)3·9H2O
Merck Index 15,4054
Solubility Information Solubility in water: soluble. Other solubilities: very soluble in ethanol, acetone
IUPAC Name iron(3+);trinitrate;nonahydrate
Formula Weight 404
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Specific Gravity 1.68
Density 1.6800g/mL
Chemical Name or Material Iron(III) nitrate nonahydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'