missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) oxalate hexahydrate, Thermo Scientific Chemicals

Product Code. 11307408
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Iron(III) oxalate hexahydrate is used as a catalyst for the reductive and oxidative degradation of iopamidol under UV and visible light irradiation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 166897-40-1
Molecular Formula C6H12Fe2O18
Molecular Weight (g/mol) 483.834
MDL Number MFCD00054430
InChI Key FWXIZVVTJVNNRX-UHFFFAOYSA-H
Synonym ferric oxalate hexahydrate
PubChem CID 131849393
IUPAC Name iron(3+);oxalate;hexahydrate
SMILES C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].O.O.O.O.O.O.[Fe+3].[Fe+3]

Specifications

Quantity 5 g
UN Number UN3288
Solubility Information Soluble in water.
Formula Weight 483.84 (375.55 Anhydrous)
Physical Form Powder
Chemical Name or Material Iron(III) oxalate hexahydrate
compliance-icons
Product Identifier
  • Iron(III) oxalate hexahydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P363-Wash contaminated clothing before reuse.
  • P321-Specific treatment (see on this label).
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.