missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(Iii) Oxide, purified, Honeywell Fluka™
Click to view available options
Quantity:
250 g
1 kg
6 x 250 g
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 250 g |
UN Number | NONH for all modes of transport |
Formula Weight | 159.69g/mol |
Percent Purity | ≥95% |
Grade | Purified |
Chemical Name or Material | Iron(Iii) Oxide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'