missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) perchlorate hydrate, Reagent Grade, Thermo Scientific Chemicals

£45.83 - £124.62

Chemical Identifiers

CAS 15201-61-3
Molecular Formula Cl3FeO12
Molecular Weight (g/mol) 354.18
MDL Number MFCD00011000
InChI Key LHOWRPZTCLUDOI-UHFFFAOYSA-K
Synonym iron iii perchlorate hydrate, perchloric acid iron iii salt hydrate, iron 3+ hydrate triperchlorate, iron iii perchlorate hydrate purple, iron iii perchlorate hydrate yellow, iron iii perchloratehydrate,reagentgrade, iron iii perchlorate hydrate, crystalline, iron iii perchlorate hydrate, crystalline, low chloride
PubChem CID 9799594
IUPAC Name iron(3+);triperchlorate;hydrate
SMILES [Fe+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11321758
SDS View Documents
Thermo Scientific Alfa Aesar
011647.22
100 g
£45.83
100g
Please to purchase this item. Need a web account? Register with us today!
11311758
SDS View Documents
Thermo Scientific Alfa Aesar
011647.36
500 g
£124.62
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Iron(III) perchlorate hydrate is used as a starting material for the synthesis of a tris-salicylate Fe(III) complex having bidentate chelating units that allow two coordination modes. It is also used as a catalyst for synthesis of fullerene-fused lactones, α-Carbonyl furans via one-pot cyclization, fullerodioxolanes via heterocyclization, dialylated indoles via double alkylation, aryl esters via oxidative esterification and as a chemical actinometer based on photolysis of ferrioxalate in presence of polyoxometalate in aqueous solution, as an oxidant for conducting polymer nanoparticles synthesized in ionic liquid by chemical polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

15201-61-3
354.18
LHOWRPZTCLUDOI-UHFFFAOYSA-K
9799594
[Fe+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Cl3FeO12
MFCD00011000
iron iii perchlorate hydrate, perchloric acid iron iii salt hydrate, iron 3+ hydrate triperchlorate, iron iii perchlorate hydrate purple, iron iii perchlorate hydrate yellow, iron iii perchloratehydrate,reagentgrade, iron iii perchlorate hydrate, crystalline, iron iii perchlorate hydrate, crystalline, low chloride
iron(3+);triperchlorate;hydrate

Specifications

15201-61-3
Crystalline
MFCD00011000
Soluble in water.
[Fe+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
354.18
354.19 (Anhydrous)
Odorless
Iron(III) perchlorate hydrate
White
Cl3FeO12
iron iii perchlorate hydrate, perchloric acid iron iii salt hydrate, iron 3+ hydrate triperchlorate, iron iii perchlorate hydrate purple, iron iii perchlorate hydrate yellow, iron iii perchloratehydrate,reagentgrade, iron iii perchlorate hydrate, crystalline, iron iii perchlorate hydrate, crystalline, low chloride
LHOWRPZTCLUDOI-UHFFFAOYSA-K
iron(3+);triperchlorate;hydrate
9799594
Reagent
Hygroscopic
Videos
Safety and Handling

Safety and Handling

P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H272-H315-H319-H335

missing translation for 'dotInformation' : Hazard Class: 5.1; Packaging Group: II

missing translation for 'einecsNumber' : 236-908-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Product Identifier
  • Iron(II) perchlorate hydrate

Signal Word
  • Danger

Hazard Category
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iron(III) perchlorate hydrate, Reagent Grade, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.