missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) phosphate hydrate, Thermo Scientific Chemicals

Product Code. 11317398
Click to view available options
Quantity:
250 g
1 kg
5 kg

Poly(methyl methacrylate) is used in the lenses of exterior lights of automobile and automotive components such as badges, steering wheel, tachometer covers, warning triangles and fascia panels. It is in the form of sheets used in shatter resistant panels for building windows, skylights, bullet proof security barriers, bathtubs, LCD screens, furniture and many other applications. It is used as the matrix in laser dye-doped organic solid-state gain media for tunable solid state dye lasers. Further, it is used as a shield to stop beta radiation emitted from radioisotopes, as dosimeter devices during the gamma irradiation process and as a dispersant for ceramic powders to stabilize colloidal suspensions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51833-68-2
Molecular Formula FeH2O5P
Molecular Weight (g/mol) 168.83
MDL Number MFCD00016096
InChI Key RQKNQWUJROKKEQ-UHFFFAOYSA-K
Synonym ferric phosphate hydrate, iron iii phosphate hydrate, ferric orthophosphate hydrate, ccris 8095, acmc-20akpj, fe.po4.h2o, ksc497s7j, iron 3+ hydrate phosphate, iron 3+ ion hydrate phosphate, phosphoric acid, iron 3 salt 1:1 , hydrate
PubChem CID 21410239
SMILES O.[Fe+3].[O-]P([O-])([O-])=O

Specifications

Color White
Physical Form Powder
Quantity 5 kg
Solubility Information Insoluble in water.
IUPAC Name iron(3+) hydrate phosphate
Formula Weight 150.82 (Anhydrous)
Appearance Off-white powder
Chemical Name or Material Iron(III) phosphate hydrate, hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'