Learn More
Iron(III) phosphate hydrate, Thermo Scientific Chemicals
£40.60 - £448.00
Chemical Identifiers
CAS | 51833-68-2 |
---|---|
Molecular Formula | FeH2O5P |
Molecular Weight (g/mol) | 168.83 |
MDL Number | MFCD00016096 |
InChI Key | RQKNQWUJROKKEQ-UHFFFAOYSA-K |
Synonym | ferric phosphate hydrate, iron iii phosphate hydrate, ferric orthophosphate hydrate, ccris 8095, acmc-20akpj, fe.po4.h2o, ksc497s7j, iron 3+ hydrate phosphate, iron 3+ ion hydrate phosphate, phosphoric acid, iron 3 salt 1:1 , hydrate |
PubChem CID | 21410239 |
SMILES | O.[Fe+3].[O-]P([O-])([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11307398
|
Thermo Scientific Alfa Aesar
031106.30 |
250 g |
£40.60
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11327398
|
Thermo Scientific Alfa Aesar
031106.A1 |
1 kg |
£105.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11317398
|
Thermo Scientific Alfa Aesar
031106.A7 |
5 kg |
£448.00
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Poly(methyl methacrylate) is used in the lenses of exterior lights of automobile and automotive components such as badges, steering wheel, tachometer covers, warning triangles and fascia panels. It is in the form of sheets used in shatter resistant panels for building windows, skylights, bullet proof security barriers, bathtubs, LCD screens, furniture and many other applications. It is used as the matrix in laser dye-doped organic solid-state gain media for tunable solid state dye lasers. Further, it is used as a shield to stop beta radiation emitted from radioisotopes, as dosimeter devices during the gamma irradiation process and as a dispersant for ceramic powders to stabilize colloidal suspensions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chemical Identifiers
51833-68-2 | |
168.83 | |
RQKNQWUJROKKEQ-UHFFFAOYSA-K | |
21410239 |
FeH2O5P | |
MFCD00016096 | |
ferric phosphate hydrate, iron iii phosphate hydrate, ferric orthophosphate hydrate, ccris 8095, acmc-20akpj, fe.po4.h2o, ksc497s7j, iron 3+ hydrate phosphate, iron 3+ ion hydrate phosphate, phosphoric acid, iron 3 salt 1:1 , hydrate | |
O.[Fe+3].[O-]P([O-])([O-])=O |
Specifications
51833-68-2 | |
Powder | |
MFCD00016096 | |
Insoluble in water. | |
O.[Fe+3].[O-]P([O-])([O-])=O | |
168.83 | |
150.82 (Anhydrous) | |
Iron(III) phosphate hydrate, hydrate |
White | |
FeH2O5P | |
ferric phosphate hydrate, iron iii phosphate hydrate, ferric orthophosphate hydrate, ccris 8095, acmc-20akpj, fe.po4.h2o, ksc497s7j, iron 3+ hydrate phosphate, iron 3+ ion hydrate phosphate, phosphoric acid, iron 3 salt 1:1 , hydrate | |
RQKNQWUJROKKEQ-UHFFFAOYSA-K | |
iron(3+);phosphate;hydrate | |
21410239 | |
Off-white powder |
Safety and Handling
H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
missing translation for 'einecsNumber' : 233-149-7
missing translation for 'rtecsNumber' : TC6190000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Iron(III) phosphate hydrate
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only