missing translation for 'onlineSavingsMsg'
Learn More
Iron lumps, vacuum remelted, low Oxygen, 99.99% (metals basis), Thermo Scientific Chemicals Product Code.: 11302048

Iron lumps, vacuum remelted, low Oxygen, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11302048
25 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11302048

Brand: Thermo Scientific Alfa Aesar 012958.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-89-6
Molecular Formula Fe
Molecular Weight (g/mol) 55.85
MDL Number MFCD00010999
InChI Key XEEYBQQBJWHFJM-UHFFFAOYSA-N
Synonym ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
PubChem CID 23925
ChEBI CHEBI:82664
IUPAC Name iron
SMILES [Fe]

Specifications

Quantity 25 g
Percent Purity 99.99%
Assay (metals basis)
Chemical Name or Material Iron lumps, vacuum remelted, Low Oxygen, Oxygen and Nitrogen analysis included

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU10K005 Certificate TypeCertificate of Analysis Date11/09/2024 Catalog Number012958.14,012958.22
Lot NumberQ02K004 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number012958.14,012958.22
Lot NumberQ02K003 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number012958.14,012958.22
Lot NumberX04J034 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number012958.14,012958.22
Lot NumberX04J035 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number012958.14,012958.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iron lumps, vacuum remelted, low Oxygen, 99.99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.