missing translation for 'onlineSavingsMsg'
Learn More

Iron Nickel powder, -325 mesh, 99.5+% (metals basis)

Product Code. 11324207
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11324207

Brand: Thermo Scientific Alfa Aesar 036683.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ni:Fe; 50:50 wt%

Iron Nickel powder is used as an low expansion alloy for precision instruments, measuring tapes and weights

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Iron Nickel powder
Color Silver to Gray
Appearance Silver gray powder
Quantity 500 g
Mesh Size -325 Mesh
Odor Odorless
Assay Percent Range >99.5% (metals basis)
Form Powder
Molecular Formula Ni:Fe; 50:50 wt%
MDL Number MFCD01741928
Solubility Information Insoluble in water.
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.