missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron powder, spherical, <20 micron, 99.5% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
250 g
1 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Specifications
Specifications
Density | 7.87g/cm3 at 20°C |
Assay Percent Range | >99.99% (metal basis), 99.5% |
Quantity | 250 g |
Form | Spherical Powder |
Chemical Name or Material | Iron |
Safety and Handling
Product Identifier
- Iron powder
- Danger
- Self-heating substance Category 1
- H251-Self-heating: may catch fire.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P235+P410-Keep cool. Protect from sunlight.
- MIXTURE LIST-Contains : Iron
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction