missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron rod, 5mm (0.2in) dia, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Form: Approx.0.155g/mm
£201.00 - £2000.00
Chemical Identifiers
CAS | 7439-89-6 |
---|---|
Molecular Formula | Fe |
Molecular Weight (g/mol) | 55.85 |
MDL Number | MFCD00010999 |
InChI Key | XEEYBQQBJWHFJM-UHFFFAOYSA-N |
Synonym | ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m |
PubChem CID | 23925 |
ChEBI | CHEBI:82664 |
IUPAC Name | iron |
SMILES | [Fe] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11311289
|
Thermo Scientific Alfa Aesar
011442.DJ |
50 mm |
£201.00
50mm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11301289
|
Thermo Scientific Alfa Aesar
011442.DO |
150 mm |
£509.00
150mm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11321289
|
Thermo Scientific Alfa Aesar
011442.HT |
5 x 150 mm |
£2000.00
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7439-89-6 | |
55.85 | |
XEEYBQQBJWHFJM-UHFFFAOYSA-N | |
23925 | |
iron |
Fe | |
MFCD00010999 | |
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m | |
CHEBI:82664 | |
[Fe] |
Specifications
Gray | |
Gray rod | |
Fe | |
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m | |
[Fe] | |
55.85 | |
CHEBI:82664 | |
Iron |
7439-89-6 | |
99.995% (Metals basis) | |
MFCD00010999 | |
XEEYBQQBJWHFJM-UHFFFAOYSA-N | |
iron | |
23925 | |
Rod |
Safety and Handling
H500
missing translation for 'einecsNumber' : 231-096-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only