missing translation for 'onlineSavingsMsg'
Learn More
Iron sponge, -50+100 mesh, 99.8% (metals basis), Thermo Scientific Chemicals Product Code.: 11354787

Iron sponge, -50+100 mesh, 99.8% (metals basis), Thermo Scientific Chemicals

Product Code. 11354787
100 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11354787

Brand: Thermo Scientific Alfa Aesar 041830.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-89-6
Molecular Formula Fe
Molecular Weight (g/mol) 55.85
MDL Number MFCD00010999
InChI Key XEEYBQQBJWHFJM-UHFFFAOYSA-N
Synonym ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
PubChem CID 23925
ChEBI CHEBI:82664
IUPAC Name iron
SMILES [Fe]

Specifications

Color Gray
Quantity 100 g
Percent Purity 99.8%
Mesh Size −50+100 Mesh
Form Sponge
Assay (metals basis)
Chemical Name or Material Iron sponge

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberC07T013 Certificate TypeCertificate of Analysis Date28/12/2021 Catalog Number041830.22,041840.K2,041830.36
Lot NumberE24M20 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number041830.22,041840.K2,041830.36
Lot NumberD15P39 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number041830.22,041840.K2,041830.36
Lot NumberE19R005 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number041830.22,041840.K2,041830.36
Lot NumberJ17T020 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number041830.22,041840.K2,041830.36
compliance-icons
Product Identifier
  • Iron sponge
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iron sponge, -50+100 mesh, 99.8% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.