missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.95% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
1 Ea.
Description
Iron sputtering target is used in semiconductor, chemical vapor deposition (CVD) and physical vapor deposition (PVD) display and optical applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Quantity | 1 Ea. |
CAS | 7439-89-6 |
Molecular Formula | Fe |
MDL Number | MFCD00010999 |
Synonym | ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m |
Solubility Information | Insoluble in water. |
InChI Key | XEEYBQQBJWHFJM-UHFFFAOYSA-N |
SMILES | [Fe] |
IUPAC Name | iron |
Molecular Weight (g/mol) | 55.85 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'