Learn More
Iron standard solution, for AAS, 1 mg/ml Fe in 2 to 5% HNO3, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 196055000
65.59 GBP valid until 2025-03-14
Use promo code "24142" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For atomic adsorption spectroscopySpecifications
Iron standard solution | |
1 mg/ml Fe in 2% HNO3 | |
Plastic bottle | |
Colorless | |
Fe | |
XEEYBQQBJWHFJM-UHFFFAOYSA-N | |
Liquid | |
iron | |
23925 | |
55.85 |
997 to 1003 μg/mL of Fe (3+) in 2% HNO3 (20°C) | |
7439-89-6, 7697-37-2, 7732-18-5 | |
500 mL | |
7.86 | |
MFCD00010999 | |
Miscible with water | |
[Fe] | |
55.85 | |
CHEBI:82664 | |
AAS |
Safety and Handling
- Iron standard solution
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H290-May be corrosive to metals.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P390-Absorb spillage to prevent material damage.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only