missing translation for 'onlineSavingsMsg'
Learn More

GE Healthcare™ Iron-supplemented Bovine Calf Serum

US Origin; Heat inactivated and irradiated.

Brand:  GE Healthcare™ SH30072.04IH

Product Code. 15725533

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Parameter: BCSS, SUP, 1000 ML, HI
  • Tested for: Bluetongue Bovine Adenovirus, Bovine Parvovirus, Bovine RSV BVDV Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
  • Sterile: Yes
  • Additional Assays: Insulin Gamma Globulin Alkaline Phosphatase Lactate Dehydrogenase SGPT SGOT pH Albumin Blood Urea Nitrogen Creatinine Total Bilirubin Sodium Potassium Calcium Chloride Inorganic Phosphorus Iron TIBC Percent Saturation Glucose IgG
  • Heat Inactivate: Yes
  • Storage: Sera should be stored at -10 ° C or lower. Once thawed, it should be stored at 2 ° to 8 ° C.
  • Sterilization Method: Filtered through sequential 100 nm pore-size rated filters.
  • Processing: Heat Inactivate
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C
Sterile
Yes
1000 mL
Bluetongue Bovine Adenovirus, Bovine Parvovirus, Bovine RSV BVDV Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers