missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTW™ Iron Test Method
Brand: WTW™ 250439
96.99 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description

Specifications
EC No. 1907/2006 | |
Iron Test Method | |
Reagent for analysis | |
3 - 4 |
|
The product is chemically stable under standard ambient conditions |
15°C to 25°C | |
Characteristic | |
14761/2 | |
0.005 to 5.00mg/L | |
Dry Powder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
WTW™ Iron Test Method > Measuring Range: 0.005 to 5.00mg/L
Spot an opportunity for improvement?Share a Content Correction