Learn More
Iron Titanium powder, -40 mesh, Thermo Scientific Chemicals
£26.90 - £70.22
Specifications
Chemical Name or Material | Iron Titanium powder |
---|---|
Color | Gray |
Avg. Mol. Wt. or Mol. Wt. Range | Fe:Ti; 60:40 wt% (Al 2% max) |
MDL Number | MFCD00198185 |
UN Number | UN3089 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Iron Titanium powder | |
Fe:Ti; 60:40 wt% (Al 2% max) | |
UN3089 | |
−40 Mesh |
Gray | |
MFCD00198185 | |
Insoluble in water. | |
Powder |
Safety and Handling
GHS H Statement
H228-H319-H335
Flammable solid.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P280-P370+P378q
H228
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Iron Titanium powder
Signal Word
- Warning
Hazard Category
- Flammable solid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H228-Flammable solid.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : Iron, Titanium
RUO – Research Use Only