Learn More
Chemsplash IRRADIATED Cape Hood Product Code.: 30058296
Estimated Shipment: 11-11-2025
to see stock.
Add to basket
Chemsplash IRRADIATED Cape Hood
GREENER_CHOICE
Product Code. 30058296
Cape Hood
Click to view available options
Cape Hood
Product Code. 30058296
Brand: Chemsplash 5775.400.999
Estimated Shipment: 11-11-2025
to see stock.
Add to basket
Add to basket
Cape Hood is made from Chemsplash soft and microporous fabric. It is elasticated at the face and covers the hair, neck and top of the shoulders.
Chemsplash Cape Hood
  • Material: 55GSM Type PB 6B Fabric
  • Sterile Irradiated and Double Bagged
  • Elasticated Facial Area
  • Cape Style Fit
  • Silicone and Latex Free
  • Anti-Static Material
  • Non-Linting
  • PFAS Free
  • Conforms to ISO 14644-1:2015 Cleanroom 5 - 9
Description: The Chemsplash Cape Hood is made from Chemsplash's soft and microporous fabric, which is strong and resistant to chemical splash and dust. This cape hood is elasticated at the face and covers the hair, neck, and top of the shoulders. It is anti-static to meet EN1149-5:2008, lint-free, and suitable for cleanroom applications. Conforms to ISO 14644-1:2015 Cleanroom 5 - 9. Chemsplash accessories can be used to protect the wearer from the environment or the environment from the wearer, ensuring maximum comfort and wearability.
Greener Choice Claims
  • Environmental benefits include:
    • Recycled Content
Specifications
Product Type Hood
Size One Size
Material Microporous
Color White
Closure Type Ties
Disposable Single-use
Latex Free Yes
Sterility Sterile
Cleanroom Class Class 100-1000 (ISO 5-6)
Product Content Correction
Product Title
Chemsplash IRRADIATED Cape Hood >