Learn More
Fisherbrand™ Irradiated Cleanroom Vinyl Tape
Irradiated vinyl cleanroom tape with low particulate and bio-burden counts that is ideal for media preparation, batching, color coding, packaging, and other applications within the lab, sterile processing and clean construction areas.
Brand:  Fisherbrand™ EGVFP-2WHIR
 View more versions of this product
Product Code. 16275162
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Irradiated vinyl cleanroom tape forms a strong bond with plastics, polymers and stainless steel without leaving adhesive residue behind.
  • Processed in a clean environment, this tape is designed for light industrial use, clean construction, and sterile processing areas.
  • Conformable with rubber adhesive.
  • Forms a strong bond with plastics, polymers, and stainless steel.
  • Does not leave adhesive residue when removed.
  • Ideal for media preparation, batching, color coding with cleanroom pen.
  • 3 inch plastic core.
Specifications
Specifications
Autoclavable
Cleanroom 1/2 inch White Vinyl Tape
Vinyl
Matte White
Vinyl
Sterile
3 in. Plastic Core
Case
40.6 x 25.4 x 15.24 cm
16°C to 27°C
36 yd.
0.5 in.
Certificate of Conformance, Certificate of Processing, NIKA - FDA, CID, Complies with Federal Specification PPP-T-66Eand CID A-A 1689A
Rubber
White
Roll
Cleanroom Tape
Sealing, Labeling, Color Coding, Floor Marking
72/Cs.
16 x 10 x 6 in.
60°F to 80°F
33 m x 1.3 cm (36 yd. x 0.5 in.)
33 m
1.3 cm