Learn More
Fisherbrand™ Irradiated Cleanroom Vinyl Tape
Irradiated vinyl cleanroom tape with low particulate and bio-burden counts that is ideal for media preparation, batching, color coding, packaging, and other applications within the lab, sterile processing and clean construction areas.
Brand:  Fisherbrand™ EGVFP-34TRIRM
 View more versions of this product
Product Code. 16285162
Description
Description
  • Irradiated vinyl cleanroom tape forms a strong bond with plastics, polymers and stainless steel without leaving adhesive residue behind.
  • Processed in a clean environment, this tape is designed for light industrial use, clean construction, and sterile processing areas.
  • Conformable with rubber adhesive.
  • Forms a strong bond with plastics, polymers, and stainless steel.
  • Does not leave adhesive residue when removed.
  • Ideal for media preparation, batching, color coding with cleanroom pen.
  • 3 inch plastic core.
Specifications
Specifications
Autoclavable
Rubber
Transparent
Roll
Cleanroom Tape
Plate sealing, marking and color coding in aseptic environments
16/Cs.
20.32 x 20.32 x 20.32 cm
16°C to 27°C
36 yd.
0.75 in.
Certificate of Conformance, Certificate of Processing, NIKA - FDA, CID, Complies with Federal Specification PPP-T-66Eand CID A-A 1689A
Vinyl
Semi-gloss
Vinyl
Sterile
3 in. Plastic Core
8 x 8 x 8 in.
60°F to 80°F
33 m x 19.05 mm (36 yd. x 0.75 in.)
33 m
19.05 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number040564-000 Certificate TypeCertificate of Quality Date27/11/2023 Catalog NumberEGVFP-34TRIRM
Lot Number026339-000 Certificate TypeCertificate of Compliance Date17/06/2021 Catalog NumberEGVFP-34TRIRM
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Irradiated Cleanroom Vinyl Tape > 3 in. core, 0.75 in. wide, transparent