Learn More
Fisherbrand™ Irradiated Cleanroom Vinyl Tape
Irradiated vinyl cleanroom tape with low particulate and bio-burden counts that is ideal for media preparation, batching, color coding, packaging, and other applications within the lab, sterile processing and clean construction areas.
Brand:  Fisherbrand™ EGVFP-34WHIRM
95.50 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.
 View more versions of this product
Product Code. 16295162
  • £109.00 / Pack of 16
Estimated Shipment: 03-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Irradiated vinyl cleanroom tape forms a strong bond with plastics, polymers and stainless steel without leaving adhesive residue behind.
  • Processed in a clean environment, this tape is designed for light industrial use, clean construction, and sterile processing areas.
  • Conformable with rubber adhesive.
  • Forms a strong bond with plastics, polymers, and stainless steel.
  • Does not leave adhesive residue when removed.
  • Ideal for media preparation, batching, color coding with cleanroom pen.
  • 3 inch plastic core.
Specifications
Specifications
Autoclavable
Rubber
White
Roll
Cleanroom Tape
Plate sealing, marking and color coding in aseptic environments
16/Cs.
20.32 x 20.32 x 20.32 cm
16°C to 27°C
36 yd.
0.75 in.
Certificate of Conformance, Certificate of Processing, NIKA - FDA, CID, Complies with Federal Specification PPP-T-66Eand CID A-A 1689A
Vinyl
Matte White
Vinyl
Sterile
3 in. Plastic Core
8 x 8 x 8 in.
60°F to 80°F
33 m x 19.05 mm (36 yd. x 0.75 in.)
33 m
19.05 mm