missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Micronova™ Hazard Bags
These low density, LDPE hazard bags are designed for containing biomedical and hazardous waste in aseptic and controlled environments.
Brand: Micronova™ BL2-25REIRBB
175.28 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.
This item is not returnable.
View return policy
Description
The Micronova Hazard Bags are constructed from FDA compliant, low density, red or yellow polyethylene and can be used with the BLSSH series Micronova BinLiner Stands. The Biohazard bags are designed for containment and disposal of biomedical waste and the Hazardous Material bags are designed for segregation and disposal of hazardous waste
- Low density polyethylene Hazard bags
- Red or Yellow Biohazard and Hazardous Material bags
- Biohazard bags are clearly marked with a Biohazard symbol in black ink
- Hazardous material bags are labeled for waste segregation in the US
- The 32YE liners work with the BLSSH-1515 BinLiner Stand
- The 24YE and 25RE work with the BLSSH-168 BinLiner Stand
- Smaller bags with a star bottom are ideal for round containers
- Larger bags are flat sealed
- Good compatibility with common cleaners
- Gamma irradiated liners are sterile for aseptic areas
Specifications
61 L | |
Five folded liners per double bag | |
15.9 in. | |
Red | |
Single-use | |
63.5 cm | |
Sterile | |
Transporting Biomedical Waste (Non-Sharps) | |
25 in. | |
36 in. | |
Hazard Bag |
Certificate of Conformance and Certificate of Irradiation Processing | |
40 cm | |
16 gal. | |
25 x 35 in. | |
91 cm | |
50/Cs. | |
Not Autoclavable | |
BLSSH-168 | |
0.050 mm | |
LDPE |