Learn More
R&D Systems™ Irradiated Mouse Embryonic Fibroblasts (5 vials)
For use as a feeder layer when culturing embryonic or induced pluripotent stem cells
Brand: R&D Systems™ PSC001
Additional Details : Weight : 0.00970kg
Description
Mitotically inactive mouse embryonic fibroblasts (MEFs) support the growth and maintenance of stem cell pluripotency by secreting necessary soluble factors and by providing a substrate for stem cell attachment.
- Ready to use
- MEFs support undifferentiated growth of embryonic and induced pluripotent stem cells
- Inactivated by gamma irradiation to avoid chemical contamination
- Consistent feeder layer to reduce experimental variation
- Free of microbial and mycoplasma contamination
Specifications
6 x 10e6 Cells/Vial, 5 Vials |