missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isatin, 98%, Thermo Scientific Chemicals
£29.60 - £122.00
Chemical Identifiers
CAS | 91-56-5 |
---|---|
Molecular Formula | C8H5NO2 |
Molecular Weight (g/mol) | 147.13 |
MDL Number | MFCD00005718 |
InChI Key | JXDYKVIHCLTXOP-UHFFFAOYSA-N |
Synonym | isatin, indoline-2,3-dione, 2,3-indolinedione, indole-2,3-dione, isatine, 2,3-dioxoindoline, pseudoisatin, tribulin, isotin, isatic acid lactam |
PubChem CID | 7054 |
ChEBI | CHEBI:27539 |
IUPAC Name | 1H-indole-2,3-dione |
SMILES | C1=CC=C2C(=C1)C(=O)C(=O)N2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10143010
|
Thermo Scientific Acros
151491000 |
100 g | Glass bottle |
£29.60
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15770809
|
Thermo Scientific Acros
|
500 g | Glass bottle |
£122.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
91-56-5 | |
147.13 | |
JXDYKVIHCLTXOP-UHFFFAOYSA-N | |
7054 | |
1H-indole-2,3-dione |
C8H5NO2 | |
MFCD00005718 | |
isatin, indoline-2,3-dione, 2,3-indolinedione, indole-2,3-dione, isatine, 2,3-dioxoindoline, pseudoisatin, tribulin, isotin, isatic acid lactam | |
CHEBI:27539 | |
C1=CC=C2C(=C1)C(=O)C(=O)N2 |
Specifications
91-56-5 | |
Orange | |
Authentic | |
C8H5NO2 | |
21, 432 | |
isatin, indoline-2,3-dione, 2,3-indolinedione, indole-2,3-dione, isatine, 2,3-dioxoindoline, pseudoisatin, tribulin, isotin, isatic acid lactam | |
JXDYKVIHCLTXOP-UHFFFAOYSA-N | |
1H-indole-2,3-dione | |
7054 | |
147.13 | |
Powder |
201°C to 204°C | |
220°C | |
98% | |
MFCD00005718 | |
15, 5148 | |
Solubility in water: 1.9g/L. Other solubilities: soluble in alkali hydroxide solutions,soluble in ethanol,diethyl ether,benzene | |
C1=CC=C2C(=C1)C(=O)C(=O)N2 | |
147.13 | |
CHEBI:27539 | |
98% | |
Isatin, 98% |
Safety and Handling
missing translation for 'einecsNumber' : 202-077-8
missing translation for 'rtecsNumber' : NL7873000
missing translation for 'tsca' : TSCA
RUO – Research Use Only