missing translation for 'onlineSavingsMsg'
Learn More

Brady™ ISO 7010 Sign - Wear ear protection

Prevent costly accidents with ISO 7010 safety signs for optimal impact and recognizability.

Brand:  Brady™

Additional Details : Weight : 0.05300kg

 View more versions of this product

Product Code. 15654447

  • £13.65 / Each
Estimated Shipment: 14-08-2024
to see stock.
Add to basket

Description

Description

  • Polypropylene
  • For indoor and outdoor use
Specifications

Specifications

ISO 7010
1 Ea.
Indoor and Outdoor
-40°C to 120°C
Global
Polypropylene
200 mm
1.501 mm
Round
White on Blue
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers