missing translation for 'onlineSavingsMsg'
Learn More

Brady™ ISO 7010 Sign - Wear eye protection

Prevent costly accidents with ISO 7010 safety signs for optimal impact and recognizability.

£7.80 - £14.55

Specifications

Certifications/Compliance ISO 7010
Material Polypropylene
Quantity 1 Ea.
For Use With (Application) Indoor and Outdoor
Thickness 1.501 mm
View More Specs

Products 2
Product Code Brand Diameter (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Price Quantity & Availability  
15634457
View Documents
Brady™
818424
100 mm
£7.80
Each
In Stock
to see stock.
Add to basket
 
15690785
View Documents
Brady™
818425
200 mm
£14.55
Each
In Stock
to see stock.
Add to basket
 
Description

Description

  • Polypropylene
  • For indoor and outdoor use
Specifications

Specifications

ISO 7010
1 Ea.
1.501 mm
Round
White on Blue
Polypropylene
Indoor and Outdoor
-40°C to 120°C
Global
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Brady™ ISO 7010 Sign - Wear eye protection

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.