missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iso-Pentane, Extra Pure, SLR, Fisher Chemical™
£253.00
Chemical Identifiers
CAS | 78-78-4 |
---|---|
Molecular Formula | C5H12 |
Molecular Weight (g/mol) | 72.15 |
MDL Number | MFCD00009338 |
InChI Key | QWTDNUCVQCZILF-UHFFFAOYSA-N |
Synonym | isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12 |
PubChem CID | 6556 |
ChEBI | CHEBI:30362 |
IUPAC Name | 2-methylbutane |
SMILES | CCC(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11914421
|
Fisher Chemical
P/1030/MS21 |
5 L | Aluminum can | N/A | |||||
Description
Iso-Pentane, Extra Pure, SLR
Chemical Identifiers
78-78-4 | |
72.15 | |
QWTDNUCVQCZILF-UHFFFAOYSA-N | |
6556 | |
2-methylbutane |
C5H12 | |
MFCD00009338 | |
isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12 | |
CHEBI:30362 | |
CCC(C)C |
Specifications
78-78-4 | |
Colorless | |
C5H12 | |
isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12 | |
CCC(C)C | |
72.15 | |
2.48 | |
72.15g/mol | |
Liquid |
-160°C | |
30°C | |
MFCD00009338 | |
QWTDNUCVQCZILF-UHFFFAOYSA-N | |
2-methylbutane | |
6556 | |
CHEBI:30362 | |
990mbar at 20°C |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 2-Methylbutane
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H411-Toxic to aquatic life with long lasting effects.
- H336-May cause drowsiness or dizziness.
- H224-Extremely flammable liquid and vapour.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /A§
- P331-Do NOT induce vomiting.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title