missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ ISO Safety Sign without Text - Warning: Biological Hazard
Safety sign for hazard identification.
£10.25
Specifications
Finish | Gloss |
---|---|
Material | Laminated Polyester |
Adhesive Type | Permanent Acrylic |
Shape | Triangular |
Color | Black on Yellow |
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
15776429
|
Brady™
139020 |
9 x 10 mm |
£10.25
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
17183833
|
Brady™
139021 |
22 x 25 mm |
£10.25
Pack of 15 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
Description
- Sign compliant to ISO 7010
- Improves workplace safety
Specifications
Gloss | |
Permanent Acrylic | |
Black on Yellow |
Laminated Polyester | |
Triangular |