missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ ISO Safety Sign without Text - Warning: Hot Surface
Safety sign for hazard identification.
£10.05 - £11.00
Specifications
Finish | Gloss |
---|---|
Material | Laminated Polyester |
Adhesive Type | Permanent Acrylic |
Shape | Triangular |
Color | Black on Yellow |
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
15629673
|
Brady™
139013 |
9 x 10 mm |
£11.00
Pack of 54 |
Estimated Shipment: 17-01-2025
Log in to see stock. |
|||||
15963057
|
Brady™
828831 |
22 x 25 mm |
£10.05
Each |
In Stock
Log in to see stock. |
|||||
Description
- Sign compliant to ISO 7010
- Improves workplace safety
Specifications
Gloss | |
Permanent Acrylic | |
Black on Yellow |
Laminated Polyester | |
Triangular |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title