missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ ISO Safety Sign without Text - Warning: Suffocation
Safety sign for hazard identification.
Brand: Brady™ 196000
2.54 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.
This item is not returnable.
View return policy
Description
- Sign compliant to ISO 7010
- Improves workplace safety
Specifications
W/W041/NT-SA-TRI100/1-B | |
Laminated Polyester | |
Triangular | |
Black on Yellow |
Gloss | |
Permanent Acrylic | |
87 x 100 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction