missing translation for 'onlineSavingsMsg'
Learn More

Brady™ ISO Vertical Safety Sign - Warning: Biological Hazard

Safety sign for hazard identification.

Brand:  Brady™ 827714

15.82 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.


Product Code. 15700059

  • £18.45 / Each
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Sign compliant to ISO 7010
  • Improves workplace safety
Specifications

Specifications

STEN W009-210x297-PP-CRD/1
Rectangular
297 x 210 mm
Black on Yellow
Polypropylene
English
Warning Biological Hazard
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.