missing translation for 'onlineSavingsMsg'
Learn More

Isoamyl acetate, 99+%, pure, Thermo Scientific Chemicals

Product Code. 10655582
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-92-2
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
MDL Number MFCD00008946
InChI Key MLFHJEHSLIIPHL-UHFFFAOYSA-N
Synonym isoamyl acetate, isopentyl acetate, isopentyl ethanoate, banana oil, pear oil, 3-methylbutyl ethanoate, isoamyl ethanoate, iso-amyl acetate, 3-methyl-1-butyl acetate, i-amyl acetate
PubChem CID 31276
ChEBI CHEBI:31725
IUPAC Name 3-methylbutyl acetate
SMILES CC(C)CCOC(C)=O

Specifications

Melting Point -78.0°C
Density 0.8740g/mL
Boiling Point 142.0°C (756.0 mmHg)
Flash Point 25°C
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3CO2CH2CH2CH(CH3)2
Refractive Index 1.3990 to 1.4010
Quantity 1 L
Beilstein 02, 132
Merck Index 15, 5156
Specific Gravity 0.874
Solubility Information Solubility in water: 2mg/mL (20°C). Other solubilities: soluble in ethanol,diethyl phthalate,mineral oil,and 1,2-propanediol
Viscosity 1.03 Pas (8.97°C)
Formula Weight 130.19
Percent Purity 99+%
Grade Pure
Chemical Name or Material Isoamyl acetate
Show More Show Less
compliance-icons
Product Identifier
  • Isoamyl acetate, mixture of 2- and 3-Methylbutyl acetate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contains : Isoamyl acetate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.