Learn More
Isoamyl Alcohol (Molecular Biology), Fisher BioReagents
Brand: Fisher Bioreagents BP1150-500
Code : 32
Additional Details : CAS Number : 123-51-3 Weight : 0.45450kg Transport : UN number : 1105 Chem class : 3 Pack group : III
Description
500mL; Glass Bottle
A mixture of Chloroform and Isoamyl Alcohol (24:1 v/v) is used to remove proteins during nucleic acid extraction procedures. It may also be used to extract Ethidium Bromide from nucleic acids.Specifications
Isoamyl Alcohol | |
0.01% max. | |
6.5 | |
≥99 % | |
C5H12O | |
500 mL | |
15, 5241 | |
PHTQWCKDNZKARW-UHFFFAOYSA-N | |
3-methylbutan-1-ol | |
31260 | |
88.15 | |
≥99% | |
Liquid |
123-51-3 | |
-117°C | |
130°C | |
Glass Bottle | |
(CH3)2CHCH2CH2OH | |
0.003% max. | |
3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol | |
CC(C)CCO | |
88.15 | |
CHEBI:15837 | |
4hPa at 20°C | |
Molecular Biology |
Safety and Handling
- 3-Methyl-1-butanol
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.