Learn More
Isoamyl nitrite, 97%, stabilized, Thermo Scientific Chemicals
£33.70 - £407.00
Chemical Identifiers
CAS | 110-46-3, 497-19-8 |
---|---|
Molecular Formula | C5H11NO2 |
Molecular Weight (g/mol) | 117.15 |
MDL Number | MFCD00002057 |
InChI Key | OWFXIOWLTKNBAP-UHFFFAOYSA-N |
Synonym | isoamyl nitrite, isopentyl nitrite, amilnitrite, vaporole, aspiral, nitrous acid, 3-methylbutyl ester, 3-methylbutanol nitrite, pentanoli nitris, nitrous acid, isopentyl ester, amyl nitrite i |
PubChem CID | 8053 |
ChEBI | CHEBI:2691 |
IUPAC Name | 3-methylbutyl nitrite |
SMILES | CC(C)CCON=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10675602
|
Thermo Scientific Acros
165701000 |
100 mL | Glass Bottle |
£33.70
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10782201
|
Thermo Scientific Acros
165705000 |
500 mL | Glass Bottle |
£105.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10669293
|
Thermo Scientific Acros
165700025 |
2.5 L | Plastic bottle |
£407.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
110-46-3, 497-19-8 | |
117.15 | |
OWFXIOWLTKNBAP-UHFFFAOYSA-N | |
8053 | |
3-methylbutyl nitrite |
C5H11NO2 | |
MFCD00002057 | |
isoamyl nitrite, isopentyl nitrite, amilnitrite, vaporole, aspiral, nitrous acid, 3-methylbutyl ester, 3-methylbutanol nitrite, pentanoli nitris, nitrous acid, isopentyl ester, amyl nitrite i | |
CHEBI:2691 | |
CC(C)CCON=O |
Specifications
110-46-3, 497-19-8 | |
0.8700g/mL | |
10°C | |
96% min. (GC) | |
(CH3)2CHCH2CH2ONO | |
MFCD00002057 | |
01,40; 04,270; 05,358; 06,307; 09,258 | |
0.87 | |
Solubility in water: practically insoluble in water. Other solubilities: miscible with alcohol,chloroform and ether | |
CC(C)CCON=O | |
0.2% sodium carbonate | |
8053 | |
117.15 | |
Liquid |
Yellow | |
96.0°C to 99.0°C | |
Authentic | |
C5H11NO2 | |
1.3840 to 1.3880 | |
01, 402 | |
15, 5168 | |
isoamyl nitrite, isopentyl nitrite, amilnitrite, vaporole, aspiral, nitrous acid, 3-methylbutyl ester, 3-methylbutanol nitrite, pentanoli nitris, nitrous acid, isopentyl ester, amyl nitrite i | |
OWFXIOWLTKNBAP-UHFFFAOYSA-N | |
3-methylbutyl nitrite | |
117.15 | |
CHEBI:2691 | |
97% | |
Isoamyl nitrite, 97% |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Highly flammable liquid and vapor.
GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-770-8
missing translation for 'rtecsNumber' : NT0187500
missing translation for 'tsca' : TSCA
- Isoamyl nitrite, stabilized
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Flammable liquid Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only