Learn More
Isoamylamine, 99%, Thermo Scientific Chemicals
£15.60 - £88.90
Chemical Identifiers
CAS | 107-85-7 |
---|---|
Molecular Formula | C5H13N |
Molecular Weight (g/mol) | 87.15 |
MDL Number | MFCD00008203 |
InChI Key | BMFVGAAISNGQNM-UHFFFAOYSA-N |
Synonym | isoamylamine, isopentylamine, 3-methylbutylamine, 1-amino-3-methylbutane, monoisoamylamine, leucamine, 1-butanamine, 3-methyl, 3-methylbutanamine, isovalerylamine, monoisopentylamine |
PubChem CID | 7894 |
ChEBI | CHEBI:43689 |
IUPAC Name | 3-methylbutan-1-amine |
SMILES | CC(C)CCN |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10612983
|
Thermo Scientific Acros
155470050 |
5 mL | Glass bottle |
£15.60
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10449070
|
Thermo Scientific Acros
155470250 |
25 mL | Glass bottle |
£32.10
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10677124
|
Thermo Scientific Acros
155471000 |
100 mL | Glass bottle |
£88.90
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
107-85-7 | |
87.15 | |
BMFVGAAISNGQNM-UHFFFAOYSA-N | |
7894 | |
3-methylbutan-1-amine |
C5H13N | |
MFCD00008203 | |
isoamylamine, isopentylamine, 3-methylbutylamine, 1-amino-3-methylbutane, monoisoamylamine, leucamine, 1-butanamine, 3-methyl, 3-methylbutanamine, isovalerylamine, monoisopentylamine | |
CHEBI:43689 | |
CC(C)CCN |
Specifications
107-85-7 | |
Colorless | |
95.0°C to 97.0°C | |
Authentic | |
C5H13N | |
1.4070 to 1.4100 | |
1106 | |
15, 5157 | |
isoamylamine, isopentylamine, 3-methylbutylamine, 1-amino-3-methylbutane, monoisoamylamine, leucamine, 1-butanamine, 3-methyl, 3-methylbutanamine, isovalerylamine, monoisopentylamine | |
BMFVGAAISNGQNM-UHFFFAOYSA-N | |
3-methylbutan-1-amine | |
7894 | |
0.6 mPa.s (20°C) | |
99% | |
Isoamylamine |
-60.0°C | |
0.7500g/mL | |
4°C | |
98.5% min. (GC) | |
(CH3)2CHCH2CH2NH2 | |
MFCD00008203 | |
04, 180 | |
0.75 | |
Solubility in water: miscible. Other solubilities: miscible with alcohol,chloroform and ether | |
CC(C)CCN | |
87.15 | |
CHEBI:43689 | |
87.15 | |
Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-526-
- Isoamylamine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only