Learn More
Isobutanol, 99%, Thermo Scientific Chemicals
£23.72 - £80.37
Chemical Identifiers
CAS | 78-83-1 |
---|---|
Molecular Formula | C4H10O |
Molecular Weight (g/mol) | 74.123 |
MDL Number | MFCD00004740 |
InChI Key | ZXEKIIBDNHEJCQ-UHFFFAOYSA-N |
Synonym | 2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol |
PubChem CID | 6560 |
ChEBI | CHEBI:46645 |
IUPAC Name | 2-methylpropan-1-ol |
SMILES | CC(C)CO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11466286
|
Thermo Scientific Alfa Aesar
B23091.AP |
500 mL |
£23.72
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11476286
|
Thermo Scientific Alfa Aesar
B23091.0F |
2500 mL |
£80.37
2500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Isobutanol is a reagent used in organic reactions. It is employed in the synthesis of new fluorinating reagents. It is also used in the lipase-catalyzed production of biodiesel as an energy source. It acts as a catalyst for the liquid-phase esterification of acetic acid by ion-exchange resins. Dehydration of fermented isobutanol is useful in the production of renewable chemicals and fuels.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIsobutanol is a reagent used in organic reactions. It is employed in the synthesis of new fluorinating reagents. It is also used in the lipase-catalyzed production of biodiesel as an energy source. It acts as a catalyst for the liquid-phase esterification of acetic acid by ion-exchange resins. Dehydration of fermented isobutanol is useful in the production of renewable chemicals and fuels.
Solubility
Miscible with alcohol and diethyl ether. Partially miscible with water.
Notes
Avoid all sources of ignition. Keep container tightly closed in a dry and well-ventilated place.
Chemical Identifiers
78-83-1 | |
74.123 | |
ZXEKIIBDNHEJCQ-UHFFFAOYSA-N | |
6560 | |
2-methylpropan-1-ol |
C4H10O | |
MFCD00004740 | |
2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol | |
CHEBI:46645 | |
CC(C)CO |
Specifications
7 | |
78-83-1 | |
107°C to 108°C | |
Sweet | |
C4H10O | |
MFCD00004740 | |
1730878 | |
2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol | |
ZXEKIIBDNHEJCQ-UHFFFAOYSA-N | |
2-methylpropan-1-ol | |
6560 | |
74.12 | |
Isobutanol |
-108°C | |
0.802 | |
28°C (82°F) | |
1.396 | |
(CH3)2CHCH2OH | |
UN1212 | |
14,5131 | |
Miscible with alcohol and diethyl ether. Partially miscible with water. | |
CC(C)CO | |
74.123 | |
CHEBI:46645 | |
99% |
Safety and Handling
GHS H Statement
H318-H226-H335-H336-H315
Causes serious eye damage.
Flammable liquid and vapour.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Causes skin irritation.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P314-P332+P313-P363-P370+P378q-P501c
H226-H315-H318-H335-H336
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ISOBUTANOL
missing translation for 'einecsNumber' : 201-148-0
missing translation for 'rtecsNumber' : NP9625000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Isobutanol
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Isobutyl alcohol
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.