missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl acetate, 98%, Thermo Scientific Chemicals

Product Code. 10667364
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Glass bottle
Metal Drum
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-19-0
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00008932
InChI Key GJRQTCIYDGXPES-UHFFFAOYSA-N
Synonym isobutyl acetate, isobutyl ethanoate, acetic acid, 2-methylpropyl ester, 2-methyl-1-propyl acetate, 2-methylpropyl ethanoate, acetic acid, isobutyl ester, acetic acid isobutyl ester, acetate d'isobutyle, iso-butyl acetate, beta-methylpropyl ethanoate
PubChem CID 8038
ChEBI CHEBI:50569
IUPAC Name 2-methylpropyl acetate
SMILES CC(C)COC(C)=O

Specifications

Melting Point -99.0°C
Density 0.8730g/mL
Boiling Point 116.0°C to 117.5°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Metal Drum
Linear Formula CH3CO2CH2CH(CH3)2
Refractive Index 1.3890 to 1.3910
Quantity 25 L
Beilstein 02, 131
Merck Index 15, 5175
Specific Gravity 0.873
Solubility Information Solubility in water: 7g/L (20°C). Other solubilities: freely soluble in alcohol,ether,propylene glycol,and other organic solvents
Viscosity 0.7 mPa.s (20°C)
Formula Weight 116.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Isobutyl acetate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.