missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl acetate, 98%, Thermo Scientific Chemicals

Product Code. 10667421
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Glass bottle
Metal Drum

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-19-0
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00008932
InChI Key GJRQTCIYDGXPES-UHFFFAOYSA-N
Synonym isobutyl acetate, isobutyl ethanoate, acetic acid, 2-methylpropyl ester, 2-methyl-1-propyl acetate, 2-methylpropyl ethanoate, acetic acid, isobutyl ester, acetic acid isobutyl ester, acetate d'isobutyle, iso-butyl acetate, beta-methylpropyl ethanoate
PubChem CID 8038
ChEBI CHEBI:50569
IUPAC Name 2-methylpropyl acetate
SMILES CC(C)COC(C)=O

Specifications

Melting Point -99.0°C
Density 0.8730g/mL
Boiling Point 116.0°C to 117.5°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CO2CH2CH(CH3)2
Refractive Index 1.3890 to 1.3910
Quantity 2.5 L
Beilstein 02, 131
Merck Index 15, 5175
Specific Gravity 0.873
Solubility Information Solubility in water: 7g/L (20°C). Other solubilities: freely soluble in alcohol,ether,propylene glycol,and other organic solvents
Viscosity 0.7 mPa.s (20°C)
Formula Weight 116.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Isobutyl acetate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'