missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl Acetate, 99%, Honeywell™

Catalog Number 15638420
Click to view available options
Quantity:
25 mL
1 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
110-19-0
C6H12O2
116.16
MFCD00008932
GJRQTCIYDGXPES-UHFFFAOYSA-N
isobutyl acetate, isobutyl ethanoate, acetic acid, 2-methylpropyl ester, 2-methyl-1-propyl acetate, 2-methylpropyl ethanoate, acetic acid, isobutyl ester, acetic acid isobutyl ester, acetate d'isobutyle, iso-butyl acetate, beta-methylpropyl ethanoate
8038
CHEBI:50569
2-methylpropyl acetate
CC(C)COC(C)=O
This item is not returnable. View return policy
110-19-0
C6H12O2
116.16
MFCD00008932
GJRQTCIYDGXPES-UHFFFAOYSA-N
isobutyl acetate, isobutyl ethanoate, acetic acid, 2-methylpropyl ester, 2-methyl-1-propyl acetate, 2-methylpropyl ethanoate, acetic acid, isobutyl ester, acetic acid isobutyl ester, acetate d'isobutyle, iso-butyl acetate, beta-methylpropyl ethanoate
8038
CHEBI:50569
2-methylpropyl acetate
CC(C)COC(C)=O

0.99

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY
Melting Point -99°C
Color Colorless
Density 0.867g/mL at 25°C
Boiling Point 117°C
Explosion Limit 0.105
Flash Point 22°C (71.6°F)
Ignition Point 793°F
Packaging Glass Bottle
Linear Formula CH3COOCH2CH(CH3)2
Refractive Index n20/D 1.39
Quantity 25 mL
UN Number UN1213
Beilstein 1741909
Vapor Density >4 (vs air)
Formula Weight 116.16g/mol
Vapor Pressure 15mmHg (20°C)
Percent Purity 99%
Chemical Name or Material Isobutyl acetate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'