missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl benzoate, 99%, Thermo Scientific™

£21.10 - £72.23

Chemical Identifiers

CAS 120-50-3
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.23
MDL Number MFCD00048344
InChI Key KYZHGEFMXZOSJN-UHFFFAOYSA-N
Synonym isobutyl benzoate, benzoic acid, isobutyl ester, benzoic acid, 2-methylpropyl ester, unii-kq6xz9wjii, isobutyl benzoate natural, fema no. 2185, kq6xz9wjii, benzoic acid isobutyl ester, benzoic acid, isobutyl ester 6ci,7ci,8ci, iso-butyl benzoate
PubChem CID 61048
ChEBI CHEBI:87500
IUPAC Name 2-methylpropyl benzoate
SMILES CC(C)COC(=O)C1=CC=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11419362
SDS View Documents
Thermo Scientific Alfa Aesar
A10114.30
250 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11409362
SDS View Documents
Thermo Scientific Alfa Aesar
A10114.0B
1000 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 204-401-3

missing translation for 'rtecsNumber' : DH4185000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Isobutyl benzoate, 99%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.