missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl propionate, 98%, Thermo Scientific™

Product Code. 15465988
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-42-1
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
MDL Number MFCD00009307
InChI Key FZXRXKLUIMKDEL-UHFFFAOYSA-N
Synonym isobutyl propionate, isobutyl propanoate, 2-methylpropyl propionate, propanoic acid, 2-methylpropyl ester, propionic acid isobutyl ester, propionic acid, isobutyl ester, methyl isobutyl acetate, 2-methyl-1-propyl propanoate, unii-8q382jsy9t, isobutyl propionate natural
PubChem CID 10895
IUPAC Name 2-methylpropyl propanoate
SMILES CCC(=O)OCC(C)C

Specifications

Melting Point -71°C
Color Colorless
Density 0.869 g/mL
Flash Point 66°C (151°F)
Linear Formula CH3CH2CO2CH2CH(CH3)2
Refractive Index 1.397
Quantity 25 mL
UN Number UN2394
Formula Weight 130.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Isobutyl propionate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'