missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isobutylamine, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mL
1000 mL
Description
Isobutylamine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Isobutylamine is used as pharmaceutical intermediate.
Solubility
Soluble in water (1000 mg/ml at 25°C), ether, alcohol, acetone and benzene.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, acids, acid chlorides and acid anhydrides.
Isobutylamine is used as pharmaceutical intermediate.
Solubility
Soluble in water (1000 mg/ml at 25°C), ether, alcohol, acetone and benzene.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, acids, acid chlorides and acid anhydrides.
Specifications
Specifications
Melting Point | -85°C |
Density | 0.736 |
Boiling Point | 67°C to 69°C |
Flash Point | −9°C (15°F) |
Assay Percent Range | 99% |
Odor | Amine-like |
Refractive Index | 1.397 |
Quantity | 1000 mL |
UN Number | UN1214 |
Beilstein | 385626 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'