Learn More
Chemical Identifiers
CAS | 78-81-9 |
---|---|
Molecular Formula | C4H11N |
Molecular Weight (g/mol) | 73.14 |
MDL Number | MFCD00008146 |
InChI Key | KDSNLYIMUZNERS-UHFFFAOYSA-N |
Synonym | isobutylamine, 1-amino-2-methylpropane, 2-methylpropylamine, monoisobutylamine, 1-propanamine, 2-methyl, valamine, iso-butylamine, i-butylamine, 2-methylpropanamine, 3-methyl-2-propylamine |
PubChem CID | 6558 |
ChEBI | CHEBI:15997 |
IUPAC Name | 2-methylpropan-1-amine |
SMILES | CC(C)CN |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10071514
|
Thermo Scientific Acros
122502500 |
250 mL | Glass bottle |
£23.52
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10366694
|
Thermo Scientific Acros
122500010 |
1 L | Glass bottle |
£61.91
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10420734
|
Thermo Scientific Acros
122500025 |
2.5 L | Glass bottle |
£136.68
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
78-81-9 | |
73.14 | |
KDSNLYIMUZNERS-UHFFFAOYSA-N | |
6558 | |
2-methylpropan-1-amine |
C4H11N | |
MFCD00008146 | |
isobutylamine, 1-amino-2-methylpropane, 2-methylpropylamine, monoisobutylamine, 1-propanamine, 2-methyl, valamine, iso-butylamine, i-butylamine, 2-methylpropanamine, 3-methyl-2-propylamine | |
CHEBI:15997 | |
CC(C)CN |
Specifications
78-81-9 | |
Colorless to Yellow | |
64.0°C to 71.0°C | |
Authentic | |
(CH3)2CHCH2NH2 | |
MFCD00008146 | |
15, 5177 | |
isobutylamine, 1-amino-2-methylpropane, 2-methylpropylamine, monoisobutylamine, 1-propanamine, 2-methyl, valamine, iso-butylamine, i-butylamine, 2-methylpropanamine, 3-methyl-2-propylamine | |
KDSNLYIMUZNERS-UHFFFAOYSA-N | |
2-methylpropan-1-amine | |
6558 | |
0.6 mPa.s (20°C) | |
≥98.5% | |
Isobutylamine |
-85.0°C | |
0.7300g/mL | |
−9°C | |
C4H11N | |
1.397 | |
04, 163 | |
0.73 | |
Solubility in water: miscible. Other solubilities: soluble in alcohol and ether | |
CC(C)CN | |
73.14 | |
CHEBI:15997 | |
73.14 | |
Liquid |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if swallowed.
GHS P Statement
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 201-145-4
- Isobutylamine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H301-Toxic if swallowed.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only